7-Methyl-1,4-dioxaspiro[4.5]decan-8-one 97%

Supplier: Ambeed
Warning

A104178-250MG A104178-5G A104178-25G
103796-666EA 38 USD
103796-666 103822-380 76869-308 77309-246 77565-814
7-Methyl-1,4-dioxaspiro[4.5]decan-8-one 97%
7-Methyl-1,4-dioxaspiro[4.5]decan-8-one

Precaución: For research use only.

Formula: C₉H₁₄O₃
Boiling Pt: 272.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD13189957
CAS Number: 702-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR