7-Methoxy-6-(3-morpholinopropoxy)quinazolin-4(3H)-one 97%

Supplier: Ambeed
Warning

Synonyms: Gefitinib impurity 5

A100552-100G A100552-10G A100552-1G A100552-5G A100552-25G
103795-508EA 8 USD
103795-508 103821-198 77025-138 77025-140 77282-508 77296-754 77666-456
7-Methoxy-6-(3-morpholinopropoxy)quinazolin-4(3H)-one 97%
7-Methoxy-6-(3-morpholinopropoxy)quinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₆H₂₁N₃O₄
MW: 319.35 g/mol
Boiling Pt: 519.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD12760130
CAS Number: 199327-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR