7-Methoxy-3,4-dihydroquinolin-2(1H)-one 98%

Supplier: Ambeed
Warning

A169197-250MG A169197-25G A169197-1G A169197-5G
103806-362EA 13 USD
103806-362 103844-616 76813-148 76813-150 77081-688 77081-690
7-Methoxy-3,4-dihydroquinolin-2(1H)-one 98%
7-Methoxy-3,4-dihydroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD15144280
CAS Number: 22246-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR