7-Methoxy-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine 95%

Supplier: Ambeed
Warning

A100788-100MG A100788-1G A100788-250MG
103799-228EA 55 USD
103799-228 76982-158 76982-160 76982-162
7-Methoxy-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine 95%
7-Methoxy-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NOS
MW: 195.28 g/mol
Boiling Pt: 336.9 °C (760 mm Hg)
Flash Pt: 157.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11112160
CAS Number: 145903-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR