7-Methoxy-1-tetralone 97%

Supplier: Ambeed
Warning

A103898-100G A103898-25G A103898-500G A103898-5G
103822-332EA 5 USD
103822-332 76765-332 76765-334 76765-336 76765-338 77309-200
7-Methoxy-1-tetralone 97%
7-Methoxy-1-tetralone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
Boiling Pt: 312.3 °C (760 mm Hg)
Melting Pt: 59…63 °C
Flash Pt: 145.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00001696
CAS Number: 6836-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR