7-Methoxy-1H-pyrrolo[3,2-b]pyridine 95%

Supplier: Ambeed
Warning

A290049-1G A290049-100MG
103813-858EA 358 USD
103813-858 103834-998 76837-578 76977-546
7-Methoxy-1H-pyrrolo[3,2-b]pyridine 95%
7-Methoxy-1H-pyrrolo[3,2-b]pyridine

Precaución: For research use only

Formula: C₈H₈N₂O
MW: 148.16 g/mol
Boiling Pt: 323.639 °C (760 mm Hg)
Flash Pt: 149.532 °C
Storage Temperature: Ambient
MDL Number: MFCD12923959
CAS Number: 425380-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR