7-Methoxy-1H-indazole 97%

Supplier: Ambeed
Warning

A202748-100MG A202748-250MG A202748-1G A202748-5G
103817-414EA 56 USD
103817-414 103848-058 77012-710 77012-712 77086-172 77302-824
7-Methoxy-1H-indazole 97%
7-Methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
Boiling Pt: 312.5 °C (760 mm Hg)
Flash Pt: 114.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08752584
CAS Number: 133841-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR