7-Methoxy-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

A367629-100MG A367629-250MG A367629-1G A367629-5G
103826-430EA 87 USD
103826-430 103837-440 76817-470 76817-472 76840-680 76840-682
7-Methoxy-1,2,3,4-tetrahydroquinoline 95%
7-Methoxy-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO
MW: 163.21 g/mol
Boiling Pt: 288.7±29.0 °C (760 mm Hg)
Flash Pt: 111.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08544271
CAS Number: 19500-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR