7-Hydroxyquinoline-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A294594-100MG
A294594-250MG
103814-014EA
332
USD
103814-014
103835-154
77166-866
77166-868
7-Hydroxyquinoline-3-carboxylic acid 95%
7-Hydroxyquinoline-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₇NO₃ MW: 189.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18417142 CAS Number: 659730-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...