Pyrazolo[1,5-a]pyrimidine-5,7-diol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Hydroxypyrazolo[1,5-a]pyrimidin-5(4H)-one
A137424-10G
A137424-25G
A137424-1G
A137424-5G
103802-454EA
9
USD
103802-454
77010-184
77010-186
77315-408
77315-410
Pyrazolo[1,5-a]pyrimidine-5,7-diol 95%
Pyrazolo[1,5-a]pyrimidine-5,7-diol
Precaución: for Research Use Only
Formula:
C₆H₅N₃O₂ MW: 151.123 g/mol Boiling Pt: 290.6 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD02091625 CAS Number: 57489-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...