7-Hydroxy-4-(trifluoromethyl)coumarin 98%

Supplier: Ambeed
Warning

A773520-250MG A773520-1G A773520-25G A773520-5G A773520-10G
103840-946EA 62 USD
103840-946 76807-330 76845-196 76845-198 76845-200 77627-538
7-Hydroxy-4-(trifluoromethyl)coumarin 98%
7-Hydroxy-4-(trifluoromethyl)coumarin

Precaución: for Research Use Only

Formula: C₁₀H₅F₃O₃
Boiling Pt: 311.4 °C (760 mm Hg)
Melting Pt: 178…180 °C
Flash Pt: 142.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00037578
CAS Number: 575-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR