7-Hydroxy-1-indanone 97%

Supplier: Ambeed
Warning

Synonyms: 7-Hydroxy-2,3-dihydro-1H-inden-1-one

A112434-10G A112434-250MG A112434-25G A112434-1G A112434-5G
103798-012EA 19 USD
103798-012 103823-846 76781-174 76781-176 76781-178 77310-650 77310-652
7-Hydroxy-1-indanone 97%
7-Hydroxy-1-indanone

Precaución: for Research Use Only

Formula: C₉H₈O₂
MW: 148.161 g/mol
Boiling Pt: 310.3 °C (760 mm Hg)
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD01548387
CAS Number: 6968-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR