7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 98%

Supplier: Ambeed
Warning

A209176-100MG A209176-250MG A209176-1G A209176-5G
103817-920EA 143 USD
103817-920 103848-606 76825-820 76825-822 77086-948 77086-950
7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde 98%
7H-Pyrrolo[2,3-d]pyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₅N₃O
MW: 147.134 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189361
CAS Number: 1060815-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR