2-Aminopurine 97%

Supplier: Ambeed
Warning

Synonyms: Purin-2-amine, 7H-Purin-2-amine

A141288-1MG A141288-250MG A141288-25G
103802-972EA 42 USD
103802-972 77010-350 77611-286 77889-951
2-Aminopurine 97%
2-Aminopurine

Precaución: for Research Use Only

Formula: C₅H₅N₅
Boiling Pt: 616.3 °C (760 mm Hg)
Melting Pt: 280…282 °C
Flash Pt: 362.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005566
CAS Number: 452-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR