7H-Dibenzo[c,g]carbazole 97%

Supplier: Ambeed
Warning

A218701-100G A218701-10G A218701-250MG A218701-25G A218701-1G A218701-5G
103818-734EA 6 USD
103818-734 103849-604 76691-718 76691-720 76691-722 76691-724 77303-506 77303-508
7H-Dibenzo[c,g]carbazole 97%
7H-Dibenzo[c,g]carbazole

Precaución: for Research Use Only

Formula: C₂₀H₁₃N
MW: 267.33 g/mol
Boiling Pt: 544.1 °C (760 mm Hg)
Melting Pt: 152…154 °C
Flash Pt: 246.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00215941
CAS Number: 194-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR