7-Fluoroisoquinoline 98%

Supplier: Ambeed
Warning

A280482-100MG A280482-250MG A280482-1G
103813-510EA 64 USD
103813-510 103853-456 76816-368 76816-370 77305-362
7-Fluoroisoquinoline 98%
7-Fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₆FN
MW: 147.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226905
CAS Number: 1075-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR