7-Fluorooxindole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Fluoroindolin-2-one
A100744-100MG
A100744-250MG
A100744-25G
103795-606EA
26
USD
103795-606
103821-304
76809-218
76809-220
77670-452
7-Fluorooxindole 97%
7-Fluorooxindole
Precaución: for Research Use Only
Formula:
C₈H₆FNO MW: 151.14 g/mol Boiling Pt: 297.9 °C (760 mm Hg) Melting Pt: 188…190 °C Flash Pt: 133.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179608 CAS Number: 71294-03-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...