7-Fluorochroman-4-one 97%

Supplier: Ambeed
Warning

Synonyms: 7-Fluoro-2,3-dihydro-4H-chromen-4-one

A126401-10G A126401-250MG A126401-1G A126401-25G A126401-5G
103800-804EA 13 USD
103800-804 76996-138 76996-140 77313-016 77313-018 77313-020
7-Fluorochroman-4-one 97%
7-Fluorochroman-4-one

Precaución: for Research Use Only

Formula: C₉H₇FO₂
MW: 166.15 g/mol
Boiling Pt: 281.9 °C (760 mm Hg)
Melting Pt: 46…50 °C
Flash Pt: 120.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06202344
CAS Number: 113209-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR