7-Fluoro-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Ambeed
Warning

A186103-100MG A186103-250MG A186103-1G
103815-912EA 187 USD
103815-912 103846-486 76974-628 76974-630 77084-134
7-Fluoro-1H-pyrrolo[3,2-c]pyridine 97%
7-Fluoro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.12 g/mol
Boiling Pt: 279.3±20.0 °C (760 mm Hg)
Flash Pt: 122.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12962591
CAS Number: 1190315-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR