7-Fluoro-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A120087-100G A120087-250MG A120087-1G
103799-878EA 18 USD
103799-878 76781-688 76781-690 77311-990
7-Fluoro-1H-indole-2-carboxylic acid 95%
7-Fluoro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.1500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02664429
CAS Number: 399-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR