7-Fluoro-1H-indazole 98%

Supplier: Ambeed
Warning

A143392-100G A143392-1G A143392-25G A143392-5G
103803-272EA 10 USD
103803-272 76689-084 77316-534 77316-536 77316-538
7-Fluoro-1H-indazole 98%
7-Fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.1264 g/mol
Boiling Pt: 274.9 °C (760 mm Hg)
Melting Pt: 123…127 °C
Flash Pt: 120.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09026976
CAS Number: 341-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR