7-Chloroquinolin-8-ol 97%

Supplier: Ambeed
Warning

A415070-1G A415070-5G A415070-100MG A415070-250MG
103827-288EA 299 USD
103827-288 103838-332 76841-904 76841-906 76877-034 76877-036
7-Chloroquinolin-8-ol 97%
7-Chloroquinolin-8-ol

Precaución: for Research Use Only

Formula: C₉H₆ClNO
Boiling Pt: 321.3 °C (760 mm Hg)
Flash Pt: 148.1 °C
Storage Temperature: Ambient
MDL Number: MFCD11870758
CAS Number: 876-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR