7-Chloro-1H-quinazoline-2,4-dione 95%

Supplier: Ambeed
Warning

A174545-100MG A174545-250MG A174545-1G A174545-5G
103806-956EA 20 USD
103806-956 103845-256 76690-266 76690-268 77082-550 77082-552
7-Chloro-1H-quinazoline-2,4-dione 95%
7-Chloro-1H-quinazoline-2,4-dione

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
Melting Pt: 360…362 °C
Storage Temperature: Ambient
MDL Number: MFCD00563867
CAS Number: 13165-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR