7-Chloropyrazolo[1,5-a]pyrimidine 98%

Supplier: Ambeed
Warning

A115744-10G A115744-250MG A115744-5G
103798-612EA 67 USD
103798-612 103824-472 76687-792 76687-794 77311-290
7-Chloropyrazolo[1,5-a]pyrimidine 98%
7-Chloropyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
Melting Pt: 96…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04035684
CAS Number: 58347-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR