7-Chloroimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A307909-10G A307909-1G A307909-5G
103814-534EA 9 USD
103814-534 103835-710 76693-536 76838-464 77305-972
7-Chloroimidazo[1,2-a]pyridine 97%
7-Chloroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD08275126
CAS Number: 4532-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR