7-Chlorobenzo[d]thiazol-2-amine 97%
Supplier: Ambeed
|
Warning
|
A156019-250MG
A156019-100MG
103804-820EA
241
USD
103804-820
103843-022
77667-202
77871-777
7-Chlorobenzo[d]thiazol-2-amine 97%
7-Chlorobenzo[d]thiazol-2-amine
Precaución: Keep in a dark place, inert atmosphere, room temperature
Formula:
C₇H₅ClN₂S Boiling Pt: 344.3 °C (760 mm Hg) Flash Pt: 162 °C Storage Temperature: Ambient |
MDL Number:
MFCD01663341 CAS Number: 20358-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...