7-Chloro-8-methylquinoline 98%

Supplier: Ambeed
Warning

A452711-5G A452711-10G
103827-808EA 19 USD
103827-808 103838-878 76842-710 77884-163
7-Chloro-8-methylquinoline 98%
7-Chloro-8-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
Boiling Pt: 278 °C (760 mm Hg)
Flash Pt: 157.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09743893
CAS Number: 78941-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR