7-Chloro-1-tetralone 98%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-3,4-dihydronaphthalen-1(2H)-one, 7-Chloro-3,4-dihydro-1(2H)-naphthalenone

A138263-10G A138263-25G A138263-5G
103802-586EA 19 USD
103802-586 77095-312 77315-602 77315-604
7-Chloro-1-tetralone 98%
7-Chloro-1-tetralone

Precaución: for Research Use Only

Formula: C₁₀H₉ClO
MW: 180.63 g/mol
Boiling Pt: 297.2 °C (760 mm Hg)
Flash Pt: 142.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01928386
CAS Number: 26673-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR