7-Chloroquinaldine 97%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-2-methylquinoline

A288315-1000G A288315-100G A288315-10G A288315-500G A288315-25G A288315-5G
103834-932EA 6 USD
103834-932 76802-334 76802-336 76802-338 76802-340 76837-492 77305-528
7-Chloroquinaldine 97%
7-Chloroquinaldine

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Boiling Pt: 278.2 °C (760 mm Hg)
Melting Pt: 74…78 °C
Flash Pt: 148.736 °C
Storage Temperature: Ambient
MDL Number: MFCD00075436
CAS Number: 4965-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR