7-Chloro-1H-pyrrolo[2,3-c]pyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Chloro-6-azaindole
A149878-100MG
A149878-10G
A149878-250MG
A149878-25G
A149878-1G
A149878-5G
103804-092EA
13
USD
103804-092
103842-218
77162-728
77162-730
77162-732
77162-734
77299-760
77317-732
7-Chloro-1H-pyrrolo[2,3-c]pyridine 98%
7-Chloro-1H-pyrrolo[2,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Boiling Pt: 335.8 °C (760 mm Hg) Flash Pt: 187.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD03839905 CAS Number: 357263-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...