7-Chloro-1H-pyrazolo[3,4-c]pyridine 97%
Supplier: Ambeed
|
Warning
|
A514900-1G
A514900-5G
A514900-10G
A514900-250MG
103828-446EA
11
USD
103828-446
103839-550
76843-540
76843-542
77034-640
77034-642
7-Chloro-1H-pyrazolo[3,4-c]pyridine 97%
7-Chloro-1H-pyrazolo[3,4-c]pyridine
Precaución: for Research Use Only
Formula:
C₆H₄ClN₃ MW: 153.5691 g/mol Boiling Pt: 357.5 °C (760 mm Hg) Melting Pt: 161…163 °C Flash Pt: 201.244 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09265710 CAS Number: 76006-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...