7-Chloroimidazo[4,5-b]pyridine 97%

Supplier: Ambeed
Warning

A153583-100MG A153583-10G A153583-250MG A153583-25G A153583-1G A153583-5G
103804-532EA 43 USD
103804-532 103842-706 76768-240 76768-242 76768-244 76768-246 77318-372 77318-374
7-Chloroimidazo[4,5-b]pyridine 97%
7-Chloroimidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
Boiling Pt: 396.5 °C (760 mm Hg)
Melting Pt: 163…167 °C
Flash Pt: 225.838 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15143399
CAS Number: 6980-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR