7-Chloro-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

A305343-1G A305343-100MG A305343-250MG
103814-428EA 59 USD
103814-428 103835-594 76838-292 76977-788 76977-790
7-Chloro-1,2,3,4-tetrahydroquinoline 95%
7-Chloro-1,2,3,4-tetrahydroquinoline

Precaución: For research use only

Formula: C₉H₁₀ClN
MW: 167.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08544268
CAS Number: 90562-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR