7-Chloro-1,2,3,4-tetrahydro-1,8-naphthyridine 95%

Supplier: Ambeed
Warning

A185652-100MG A185652-250MG A185652-1G
103815-872EA 227 USD
103815-872 103846-442 77012-096 77012-098 77301-990
7-Chloro-1,2,3,4-tetrahydro-1,8-naphthyridine 95%
7-Chloro-1,2,3,4-tetrahydro-1,8-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₉ClN₂
MW: 168.62 g/mol
Boiling Pt: 372.8±34.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD20688598
CAS Number: 1303588-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR