7-Bromoquinoxalin-2(1H)-one 95%

Supplier: Ambeed
Warning

A284838-10G A284838-250MG A284838-1G A284838-25G A284838-5G
103813-654EA 18 USD
103813-654 103834-788 76693-122 76693-124 76837-314 76837-316 76837-318
7-Bromoquinoxalin-2(1H)-one 95%
7-Bromoquinoxalin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MW: 225.0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01465145
CAS Number: 82031-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR