7-Bromoquinolin-2-amine 95%

Supplier: Ambeed
Warning

A265777-100MG A265777-250MG
103812-754EA 179 USD
103812-754 103852-650 76988-246 77612-546
7-Bromoquinolin-2-amine 95%
7-Bromoquinolin-2-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062656
CAS Number: 116632-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR