7-Bromoquinazoline-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A161683-250MG A161683-5G
103805-516EA 11 USD
103805-516 103843-756 77028-650 77319-746
7-Bromoquinazoline-2,4(1H,3H)-dione 98%
7-Bromoquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954848
CAS Number: 114703-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR