7-Bromoquinazoline 97%

Supplier: Ambeed
Warning

A189814-1G A189814-10G A189814-250MG
103816-222EA 44 USD
103816-222 103846-818 77084-590 77111-710 77111-712
7-Bromoquinazoline 97%
7-Bromoquinazoline

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
MW: 209.0400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778500
CAS Number: 89892-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR