4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromopyrrolo[2,1-f][1,2,4]triazin-4-amine

A632729-10G A632729-250MG A632729-25G A632729-1G A632729-5G
103840-286EA 26 USD
103840-286 76819-628 76819-630 76819-632 76844-462 76844-464
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine 97%
4-Amino-7-bromo-pyrrolo[2,1-f][1,2,4]triazine

Precaución: for Research Use Only

Formula: C₆H₅BrN₄
MW: 213.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09033848
CAS Number: 937046-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR