7-Bromo-2-naphthol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Bromonaphthalen-2-ol, 7-Bromo-2-hydroxynaphthalene, 2-Bromo-7-hydroxynaphthalene
A113024-100G
A113024-10G
A113024-25G
A113024-500G
A113024-1G
A113024-5G
103798-146EA
6
USD
103798-146
103823-982
77148-794
77148-796
77148-798
77148-800
77310-818
77310-820
7-Bromo-2-naphthol 98%
7-Bromo-2-naphthol
Precaución: for Research Use Only
Formula:
C₁₀H₇BrO MW: 223.07 g/mol Melting Pt: 133…138 °C Flash Pt: 167.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00277644 CAS Number: 116230-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...