7-Bromoisoquinoline 97%

Supplier: Ambeed
Warning

A142957-100G A142957-100MG A142957-250MG A142957-1G A142957-10G
103803-228EA 13 USD
103803-228 77109-880 77109-882 77109-884 77316-472 77566-866
7-Bromoisoquinoline 97%
7-Bromoisoquinoline

Precaución: For research use only.

Formula: C₉H₆BrN
MW: 208.0600 g/mol
Melting Pt: 67…72 °C
Flash Pt: 142.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07368661
CAS Number: 58794-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR