7-Bromoisoquinolin-1-ol 96%

Supplier: Ambeed
Warning

A105736-10G A105736-250MG A105736-25G A105736-1G A105736-5G
103796-930EA 45 USD
103796-930 103822-670 77257-418 77257-420 77257-422 77309-534 77309-536
7-Bromoisoquinolin-1-ol 96%
7-Bromoisoquinolin-1-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Boiling Pt: 427.5 °C (760 mm Hg)
Melting Pt: 248…250 °C
Storage Temperature: Ambient
MDL Number: MFCD01861851
CAS Number: 223671-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR