7-Bromoindole 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-1H-indole

A211565-100G A211565-1G A211565-5G
103818-114EA 17 USD
103818-114 103848-808 76770-526 77087-220 77087-222
7-Bromoindole 97%
7-Bromoindole

Precaución: for Research Use Only

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 316.9 °C (760 mm Hg)
Melting Pt: 41…44 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00799492
CAS Number: 51417-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR