7-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A269658-100MG A269658-250MG
103812-972EA 90 USD
103812-972 103852-878 76827-294 76827-296
7-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde 98%
7-Bromoimidazo[1,2-a]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MW: 225.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09994348
CAS Number: 1019020-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR