7-Bromoimidazo[1,2-a]pyridine 95%

Supplier: Ambeed
Warning

A110482-250MG A110482-1G
103797-670EA 8 USD
103797-670 103823-476 77025-744 77310-270
7-Bromoimidazo[1,2-a]pyridine 95%
7-Bromoimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778353
CAS Number: 808744-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR