7-Bromoheptanoic acid 97%
Supplier: Ambeed
|
Warning
|
A115404-10G
A115404-250MG
A115404-25G
A115404-1G
A115404-5G
A115404-100G
A115404-500G
103798-558EA
6
USD
103798-558
103824-414
76995-492
76995-494
76995-496
77311-230
77311-232
77592-246
77670-620
7-Bromoheptanoic acid 97%
7-Bromoheptanoic acid
Precaución: for Research Use Only
Formula:
C₇H₁₃BrO₂ Boiling Pt: 280.7 °C (760 mm Hg) Melting Pt: 136…138 °C Flash Pt: 123.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037800 CAS Number: 30515-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...