7-Bromofuro[3,2-c]pyridin-4(5H)-one 98%

Supplier: Ambeed
Warning

A179876-100MG A179876-250MG A179876-1G
103807-484EA 144 USD
103807-484 103845-838 76872-704 76872-706 77083-314
7-Bromofuro[3,2-c]pyridin-4(5H)-one 98%
7-Bromofuro[3,2-c]pyridin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₄BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD11846190
CAS Number: 603301-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR