7-Bromobenzo[b]furan 98%

Supplier: Ambeed
Warning

Synonyms: 7-Bromobenzofuran

A118545-250MG A118545-25G A118545-1G A118545-10G
103799-072EA 10 USD
103799-072 77026-258 77026-260 77311-742 77571-102
7-Bromobenzo[b]furan 98%
7-Bromobenzo[b]furan

Precaución: for Research Use Only

Formula: C₈H₅BrO
MW: 197.031 g/mol
Boiling Pt: 233.8 °C (760 mm Hg)
Flash Pt: 95.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09056756
CAS Number: 133720-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR