7-Bromobenzo[d]thiazole 98%

Supplier: Ambeed
Warning

A482725-100MG A482725-250MG A482725-1G A482725-5G
103828-146EA 33 USD
103828-146 103839-220 76789-700 76789-702 76843-162 76843-164
7-Bromobenzo[d]thiazole 98%
7-Bromobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₄BrNS
MW: 214.0824 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11858589
CAS Number: 767-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR