7-Bromobenzo[d]thiazol-2(3H)-one 95%

Supplier: Ambeed
Warning

A203394-100MG A203394-250MG A203394-1G A203394-5G
103817-460EA 707 USD
103817-460 103848-114 76825-624 76825-626 77302-846 77302-848
7-Bromobenzo[d]thiazol-2(3H)-one 95%
7-Bromobenzo[d]thiazol-2(3H)-one

Precaución: for Research Use Only

Formula: C₇H₄BrNOS
Storage Temperature: Ambient
MDL Number: MFCD09749232
CAS Number: 1188047-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR